Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

640492

Sigma-Aldrich

2-Nitrobenzenesulfenyl chloride-13C6

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2N13C6H4SCl
CAS Number:
Molecular Weight:
195.58
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

assay

98% (CP)

technique(s)

protein expression: suitable

mp

72-75 °C (lit.)

mass shift

M+6

storage temp.

2-8°C

SMILES string

[O-][N+](=O)[13c]1[13cH][13cH][13cH][13cH][13c]1SCl

InChI

1S/C6H4ClNO2S/c7-11-6-4-2-1-3-5(6)8(9)10/h1-4H/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

NTNKNFHIAFDCSJ-IDEBNGHGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service