Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

637203

Sigma-Aldrich

3,4-Ethylenedioxypyrrole-2,5-dicarboxylic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NO6
CAS Number:
Molecular Weight:
213.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

95%

mp

137-154 °C (lit.)

SMILES string

OC(=O)c1[nH]c(C(O)=O)c2OCCOc12

Application

Starting monomer for a variety of etheylenedioxypyrrole (EDOP) conductive and electrooptical polymers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Schottland, P.; Reynolds, J.;
Macromolecules, 33, 7051-7051 (2000)
K Zong et al.
The Journal of organic chemistry, 66(21), 6873-6882 (2001-10-13)
New functionalized derivatives of 3,4-ethylenedioxypyrrole (EDOP, 5a) and 3,4-(1,3-propylenedioxy)pyrrole (ProDOP, 5b) as especially electron-rich monomers which yield highly electroactive and stable conducting polymers useful for a diverse set of applications have been synthesized. N-Alkylations of ProDOP were carried out to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service