633143
3-(1,4-Dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenylmagnesium bromide solution
0.25 M in THF
About This Item
concentration
0.25 M in THF
density
0.925 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
Br[Mg]c1cccc(CN2CCC3(CC2)OCCO3)c1
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3
target_organs
Respiratory system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
1.4 °F
flash_point_c
-17 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service