Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

632856

Sigma-Aldrich

4-Bromo-2,5-difluorobenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$31.16

$31.16

List Price$32.80
Web-Only Promotion

Available to ship onApril 22, 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$31.16

About This Item

Empirical Formula (Hill Notation):
C6H2BrClF2O2S
CAS Number:
Molecular Weight:
291.50
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$31.16

List Price$32.80
Web-Only Promotion

Available to ship onApril 22, 2025Details


Request a Bulk Order

Quality Level

assay

97%

form

solid

mp

38-42 °C (lit.)

functional group

bromo
fluoro

SMILES string

Fc1cc(c(F)cc1Br)S(Cl)(=O)=O

InChI

1S/C6H2BrClF2O2S/c7-3-1-5(10)6(2-4(3)9)13(8,11)12/h1-2H

InChI key

SBMKFWMFNIEPDN-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service