Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

631582

Sigma-Aldrich

(R)-(+)-2,2′-Bis(methoxymethoxy)-1,1′-binaphthalene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OCH2OC10H6)2
CAS Number:
Molecular Weight:
374.43
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

optical activity

[α]20/D +92°, c = 1% in chloroform

mp

101-105 °C (lit.)

SMILES string

COCOc1ccc2ccccc2c1-c3c(OCOC)ccc4ccccc34

InChI

1S/C24H22O4/c1-25-15-27-21-13-11-17-7-3-5-9-19(17)23(21)24-20-10-6-4-8-18(20)12-14-22(24)28-16-26-2/h3-14H,15-16H2,1-2H3

InChI key

YIAQRNNJNMLGTP-UHFFFAOYSA-N

Application

3- or 3,3′-di-substituted products can be obtained easily via ortho-metallation and subsequent treatment with an electrophile to give the corresponding BINOL derivatives.[1]

pictograms

CorrosionEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cox, P. J. et al
Tetrahedron Letters, 33, 2253-2253 (1992)

Articles

We present an article concerning BINOL and Derivatives.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service