631167
2-Bromo-N-methylbenzylamine
97%
Synonym(s):
(2-Bromobenzyl)methylamine
About This Item
Recommended Products
assay
97%
refractive index
n20/D 1.5670 (lit.)
bp
71-74 °C/2 mmHg (lit.)
density
1.358 g/mL at 25 °C (lit.)
SMILES string
CNCc1ccccc1Br
InChI
1S/C8H10BrN/c1-10-6-7-4-2-3-5-8(7)9/h2-5,10H,6H2,1H3
InChI key
TUADRPBKJHMHDH-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
219.9 °F - closed cup
flash_point_c
104.4 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service