Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

631132

Sigma-Aldrich

3-Methyl-N-methylbenzylamine

97%

Synonym(s):

(3-Methylbenzyl)methylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N
CAS Number:
Molecular Weight:
135.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.5190 (lit.)

bp

90-92 °C/15 mmHg (lit.)

density

0.900 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CNCc1cccc(C)c1

InChI

1S/C9H13N/c1-8-4-3-5-9(6-8)7-10-2/h3-6,10H,7H2,1-2H3

InChI key

WCTNVGNEUDTSOZ-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

174.0 °F - closed cup

flash_point_c

78.9 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryce J Marquis et al.
Chromatographia, 80(12), 1723-1732 (2017-12-08)
Small molecules containing carboxylic acid functional groups are ubiquitous throughout biology, playing vital roles in biological chemistry ranging from energy metabolism to cellular signaling. This paper describes a new derivatization reagent, 4-bromo-

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service