Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

630519

Sigma-Aldrich

5-Hexyl-2,2′-bithiophene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18S2
CAS Number:
Molecular Weight:
250.42
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

refractive index

n20/D 1.5890 (lit.)

bp

355 °C (lit.)

density

1.060 g/mL at 25 °C (lit.)

SMILES string

CCCCCCc1ccc(s1)-c2cccs2

InChI

1S/C14H18S2/c1-2-3-4-5-7-12-9-10-14(16-12)13-8-6-11-15-13/h6,8-11H,2-5,7H2,1H3

InChI key

NUONHINJVGHSCL-UHFFFAOYSA-N

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - Eye Dam. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service