Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60784

Sigma-Aldrich

Cobalt

puriss. p.a., ≥99.8%, powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

puriss. p.a.

assay

≥99.8%

form

powder

reaction suitability

reagent type: catalyst
core: cobalt

resistivity

6.24 μΩ-cm, 20°C

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

cation traces

Ca: ≤100 mg/kg
Cd: ≤10 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤100 mg/kg
K: ≤100 mg/kg
Mg: ≤50 mg/kg
Mn: ≤10 mg/kg
Na: ≤100 mg/kg
Ni: ≤500 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameHealth hazard

signalword

Danger

Hazard Classifications

Aquatic Chronic 4 - Carc. 1B - Eye Irrit. 2 - Flam. Sol. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dilemmas about the toxicological consequences of metal-on-metal hip prostheses -- What we do and do not know, and what we should do?
Jeffrey Brent et al.
Clinical toxicology (Philadelphia, Pa.), 51(4), 195-198 (2013-04-06)
Jeannette Ø Penny et al.
Acta orthopaedica, 84(2), 130-137 (2013-04-20)
Wear particles from metal-on-metal arthroplasties are under suspicion for adverse effects both locally and systemically, and the DePuy ASR Hip Resurfacing System (RHA) has above-average failure rates. We compared lymphocyte counts in RHA and total hip arthroplasty (THA) and investigated
S Perconti et al.
Journal of biological regulators and homeostatic agents, 27(2), 443-454 (2013-07-09)
Size-dependent characteristics of novel engineered nanomaterials might result in unforeseen biological responses and toxicity. To address this issue, we used cDNA microarray analysis (13443 genes) coupled with bioinformatics and functional gene annotation studies to investigate the transcriptional profiles of Balb/3T3
Takuya Kurahashi et al.
Inorganic chemistry, 52(7), 3908-3919 (2013-03-23)
The Co(salen)(X) complex, where salen is chiral N,N'-bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine and X is an external axial ligand, has been widely utilized as a versatile catalyst. The Co(salen)(X) complex is a stable solid that has been conventionally described as a Co(III)(salen)(X) complex. Recent
Chuanyu Sun et al.
Journal of nanoscience and nanotechnology, 13(1), 236-241 (2013-05-08)
Owing to their great potentialities of carbon nanotubes (CNTs)-based magnetic nano-composites, numerous applications of them have been found in nanotechnology, integrated functional system, and in medicine. Herein, nearly monodisperse CoFe2O4 nanoparticles have been deposited on multi-walled carbon nanotubes (MWCNTs) by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service