Skip to Content
MilliporeSigma
All Photos(1)

Documents

588792

Sigma-Aldrich

Boc-Asp-OH-15N

98 atom % 15N

Synonym(s):

N-(tert-Butoxycarbonyl)-L-aspartic acid-15N, L-Aspartic-15N acid, N-t-Boc derivative

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(15NH-Boc)CO2H
CAS Number:
Molecular Weight:
234.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

98 atom % 15N

form

solid

optical activity

[α]20/D -7.3°, c = 2 in DMF

technique(s)

bio NMR: suitable

mp

116-118 °C (lit.)

application(s)

peptide synthesis

functional group

Boc

mass shift

M+1

SMILES string

CC(C)(C)OC(=O)[15NH][C@@H](CC(O)=O)C(O)=O

InChI

1S/C9H15NO6/c1-9(2,3)16-8(15)10-5(7(13)14)4-6(11)12/h5H,4H2,1-3H3,(H,10,15)(H,11,12)(H,13,14)/t5-/m0/s1/i10+1

InChI key

KAJBMCZQVSQJDE-PYWYPUIGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service