Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

588385

Sigma-Aldrich

Butane-1,1,1-d3

98 atom % D

Synonym(s):

1,1,1-Trideuterobutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)2CD3
CAS Number:
Molecular Weight:
61.14
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

bp

-0.5 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])CCC

InChI

1S/C4H10/c1-3-4-2/h3-4H2,1-2H3/i1D3

InChI key

IJDNQMDRQITEOD-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

FlameGas cylinder

signalword

Danger

hcodes

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class

2A - Gases

wgk_germany

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service