Skip to Content
MilliporeSigma
All Photos(1)

Documents

587753

Sigma-Aldrich

Hexachloroethane-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C13CCl3
CAS Number:
Molecular Weight:
237.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

99 atom % 13C

mp

190-195 °C (subl.) (lit.)

mass shift

M+1

SMILES string

ClC(Cl)(Cl)[13C](Cl)(Cl)Cl

InChI

1S/C2Cl6/c3-1(4,5)2(6,7)8/i1+1

InChI key

VHHHONWQHHHLTI-OUBTZVSYSA-N

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service