Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

586080

Sigma-Aldrich

Ethylamine-N,N-d2

99 atom % D

Synonym(s):

N,N-Dideuterioethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2ND2
CAS Number:
Molecular Weight:
47.10
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % D

bp

16.6 °C (lit.)

mass shift

M+2

SMILES string

[2H]N([2H])CC

InChI

1S/C2H7N/c1-2-3/h2-3H2,1H3/i/hD2

Inchi Key

QUSNBJAOOMFDIB-ZSJDYOACSA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Gas 1A - Press. Gas Liquefied gas - STOT SE 3

target_organs

Respiratory system

Storage Class

2A - Gases

wgk_germany

WGK 1

flash_point_f

1.4 °F - closed cup

flash_point_c

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service