Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

570184

Sigma-Aldrich

2-Acetoxyphenylboronic acid pinacol ester

Synonym(s):

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4OCOCH3BO2C2(CH3)4
CAS Number:
Molecular Weight:
262.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

47-51 °C (lit.)

functional group

ester

SMILES string

CC(=O)Oc1ccccc1B2OC(C)(C)C(C)(C)O2

InChI

1S/C14H19BO4/c1-10(16)17-12-9-7-6-8-11(12)15-18-13(2,3)14(4,5)19-15/h6-9H,1-5H3

InChI key

KFBOKXXUJHAZIH-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service