Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

569003

Sigma-Aldrich

2-Bromo-4-methylphenyl isocyanate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3BrNCO
CAS Number:
Molecular Weight:
212.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.5770 (lit.)

bp

215-216 °C (lit.)

density

1.494 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(N=C=O)c(Br)c1

InChI

1S/C8H6BrNO/c1-6-2-3-8(10-5-11)7(9)4-6/h2-4H,1H3

InChI key

HUSGEXYNVCQXKA-UHFFFAOYSA-N

Caution

Refrigerate

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service