Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

567612

Sigma-Aldrich

3,4,5-Trimethoxybenzamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H3CO)3C6H2C(O)NH2
CAS Number:
Molecular Weight:
211.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

97%

mp

179-184 °C (lit.)

SMILES string

COc1cc(cc(OC)c1OC)C(N)=O

InChI

1S/C10H13NO4/c1-13-7-4-6(10(11)12)5-8(14-2)9(7)15-3/h4-5H,1-3H3,(H2,11,12)

Inchi Key

GGNMTJKRHHLJHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Eckers et al.
European journal of mass spectrometry (Chichester, England), 11(1), 73-82 (2005-06-11)
This work describes electrospray ionisation tandem mass spectrometry studies of trimethoprim and a series of structurally similar compounds containing alkoxy-phenyl groups; using accurate mass measurement to confirm the proposed fragmentations. Radical cations were observed in the spectra obtained for some
[Studies on antiarrhythmics. I. Synthesis of 3,4,5,-trimethoxybenzamide derivatives (author's transl)].
Y S Liu et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 16(2), 158-160 (1981-02-01)
Xianfu Lu et al.
Journal of neuroscience methods, 168(1), 98-103 (2008-01-09)
The distal cerebrospinal fluid-contacting neurons (dCSF-CNs) are one special type of neurons, whose bodies are in the parenchyma of the brain and processes extend into the ventricle, which suggested that dCSF-CNs might play the important roles in neuromodulation and neuroendocrinal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service