567124
1,2-Bis(diphenylphosphino)ethane monooxide
95%
Synonym(s):
[2-(Diphenylphosphino)ethyl]diphenylphosphine oxide
About This Item
Recommended Products
assay
95%
form
solid
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
mp
195-199 °C (lit.)
functional group
phosphine
SMILES string
O=P(CCP(c1ccccc1)c2ccccc2)(c3ccccc3)c4ccccc4
InChI
1S/C26H24OP2/c27-29(25-17-9-3-10-18-25,26-19-11-4-12-20-26)22-21-28(23-13-5-1-6-14-23)24-15-7-2-8-16-24/h1-20H,21-22H2
InChI key
JCAUAUKUBUBNSB-UHFFFAOYSA-N
Application
- Radical polymerization of methacrylates
- Asymmetric allylic alkylations
- Hydroformylation of dihydromyrcenol and epoxides
Linker having an effect on biodistribution of PET tumor imaging agents
Reactant for Wittig-Horner olefination of aldehydes
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service