Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

563617

Sigma-Aldrich

3,5-Dichlorobenzenesulfonamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3Cl2SO2NH2
CAS Number:
Molecular Weight:
226.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

mp

161-165 °C (lit.)

SMILES string

NS(=O)(=O)c1cc(Cl)cc(Cl)c1

InChI

1S/C6H5Cl2NO2S/c7-4-1-5(8)3-6(2-4)12(9,10)11/h1-3H,(H2,9,10,11)

InChI key

AHNOVNYOUPQVRX-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Ryan et al.
British journal of pharmacology, 174(14), 2209-2224 (2017-04-06)
With the emergence of extensively drug-resistant tuberculosis, there is a need for new anti-tubercular drugs that work through novel mechanisms of action. The meta cleavage product hydrolase, HsaD, has been demonstrated to be critical for the survival of Mycobacterium tuberculosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service