Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

563277

Sigma-Aldrich

4-Isopropylbenzyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4CH(CH3)2CH2Cl
CAS Number:
Molecular Weight:
168.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

refractive index

n20/D 1.5230 (lit.)

density

1.5230 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

CC(C)c1ccc(CCl)cc1

InChI

1S/C10H13Cl/c1-8(2)10-5-3-9(7-11)4-6-10/h3-6,8H,7H2,1-2H3

InChI key

CYAKWEQUWJAHLW-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service