About This Item
Recommended Products
assay
96%
refractive index
n20/D >1.5890 (lit.)
density
1.576 g/mL at 25 °C (lit.)
SMILES string
Clc1ccc(cc1Cl)C(Cl)(Cl)Cl
InChI
1S/C7H3Cl5/c8-5-2-1-4(3-6(5)9)7(10,11)12/h1-3H
InChI key
ATYLRBXENHNROH-UHFFFAOYSA-N
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service