Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56146

Sigma-Aldrich

3-[Hydroxy(2-phenyl-1,3-dithian-2-yl)methyl]phenoxymethyl-polystyrene

extent of labeling: ~0.6 mmol/g loading

Synonym(s):

safety-catch on polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

extent of labeling

~0.6 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

100-300 mesh

Other Notes

This safety-catch photolabile resin for solid phase organic syntheses cannot undergo premature photolysis prior to dithiane hydrolysis[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Routledge et al.
Tetrahedron Letters, 38, 1227-1227 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service