Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

558745

Sigma-Aldrich

4-Bromo-2,6-dichlorobenzenesulfonyl chloride

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2BrCl3O2S
CAS Number:
Molecular Weight:
324.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

solid

mp

37-42 °C (lit.)

SMILES string

Clc1cc(Br)cc(Cl)c1S(Cl)(=O)=O

InChI

1S/C6H2BrCl3O2S/c7-3-1-4(8)6(5(9)2-3)13(10,11)12/h1-2H

InChI key

CKJIKXAPXLPSCL-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service