Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

556297

Sigma-Aldrich

2,3-Dichloro-N-phenylmaleimide

Synonym(s):

3,4-Dichloro-1-phenyl-pyrrol-2,5-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5Cl2NO2
CAS Number:
Molecular Weight:
242.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

mp

199-205 °C (lit.)

functional group

chloro
imide
maleimide

SMILES string

ClC1=C(Cl)C(=O)N(c2ccccc2)C1=O

InChI

1S/C10H5Cl2NO2/c11-7-8(12)10(15)13(9(7)14)6-4-2-1-3-5-6/h1-5H

InChI key

CZLKFADFZAWWBC-UHFFFAOYSA-N

General description

2,3-Dichloro-N-phenylmaleimide can be prepared from the reaction between 2,3-dichloromaleic anhydride and aniline.[1]

Application

2,3-Dichloro-N-phenylmaleimide may be used to synthesize:
  • N-phenyl-2,3-di(thiophenyl)maleimide[1]
  • N-phenyl-2,3-dithiomaleimide[1]
  • 2-chloro-3-cyclohexyl-N-phenylmaleimide[2]
  • 2,3-dicyclohexyl-N-phenylmaleimide[2]
  • 2-benzyl-3-chloro-N-phenylmaleimide[2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and properties of N-phenylmaleimide derivatives.
Bodige SG, et al.
Journal of Chemical Crystallography, 29(1), 57-66 (1999)
A facile synthesis of alkyl substituted maleic anhydrides: radical approach.
Messorosh AV, et al.
Tetrahedron, 64(48), 10849-10852 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service