Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

553581

Sigma-Aldrich

6-Bromo-2-hydroxy-3-methoxybenzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(OH)(OCH3)CHO
CAS Number:
Molecular Weight:
231.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

102-105 °C (lit.)

SMILES string

COc1ccc(Br)c(C=O)c1O

InChI

1S/C8H7BrO3/c1-12-7-3-2-6(9)5(4-10)8(7)11/h2-4,11H,1H3

InChI key

JUPJZUTYDWXZAQ-UHFFFAOYSA-N

General description

6-Bromo-2-hydroxy-3-methoxybenzaldehyde is a bromobenzaldehyde derivative.[1] It participates in the synthesis of (±)-norannuradhapurine.[2] Its crystals exhibit monoclinic crystal system and space group P21/n.[1]

Application

6-Bromo-2-hydroxy-3-methoxybenzaldehyde may be used in the synthesis of:
  • 2-benzyloxy-6-bromo-3-methoxybenzaldehyde[3]
  • biphenyls[4]
  • antihypertensive natural products S-(+)-XJP and R-(-)-XJP[5]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic studies on selective adenosine A 2A receptor antagonists: Synthesis and structure-activity relationships of novel benzofuran derivatives
Saku O, et al.
Bioorganic & Medicinal Chemistry Letters, 20(3), 1090-1093 (2010)
Total synthesis and the biological activities of (?)-norannuradhapurine.
Nimgirawath S, et al.
Molecules (Basel), 14(1), 89-101 (2008)
Synthesis of NK109, an anticancer benzo [c] phenanthridine alkaloid.
Nakanishi T, et al.
The Journal of Organic Chemistry, 63(13), 4235-4239 (1998)
The conformation of some ortho-bromoarylaldehydes.
Hanson JR, et al.
J. Chem. Res. (M), 7, 416-418 (2008)
First total synthesis of antihypertensive natural products S-(+)-XJP and R-(-)-XJP.
Wang C, et al.
Organic & Biomolecular Chemistry, 12(37), 7338-7344 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service