Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

553174

Sigma-Aldrich

3,4-Dihydroxythiophene-2,5-dicarboxylic acid diethyl ester

Synonym(s):

2,5-Dicarbethoxy-3,4-dihydroxythiophene, 2,5-Diethoxycarbonyl-3,4-dihydroxythiophene, 3,4-Dihydroxy-2,5-dicarbethoxythiophene, 3,4-Dihydroxythiophene-2,5-dicarboxylic acid diethyl ester, Dicetol, Diethyl 3,4-dihydroxy-2,5-thiophenedicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O6S
CAS Number:
Molecular Weight:
260.26
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

mp

132-135 °C (lit.)

SMILES string

CCOC(=O)c1sc(c(O)c1O)C(=O)OCC

InChI

1S/C10H12O6S/c1-3-15-9(13)7-5(11)6(12)8(17-7)10(14)16-4-2/h11-12H,3-4H2,1-2H3

InChI key

YOKOBAHUBJPMGP-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hirokazu Yano et al.
Science advances, 5(4), eaav9492-eaav9492 (2019-04-18)
Wet-processable and highly conductive polymers are promising candidates for key materials in organic electronics. Poly(3,4-ethylenedioxythiophene):poly(4-styrenesulfonate) (PEDOT:PSS) is commercially available as a water dispersion of colloidal particles but has some technical issues with PSS. Here, we developed a novel fully soluble

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service