Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

549266

Sigma-Aldrich

3,5-Dichlorothioanisole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3SCH3
CAS Number:
Molecular Weight:
193.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.613 (lit.)

bp

262-263 °C (lit.)

density

1.349 g/mL at 25 °C (lit.)

SMILES string

CSc1cc(Cl)cc(Cl)c1

InChI

1S/C7H6Cl2S/c1-10-7-3-5(8)2-6(9)4-7/h2-4H,1H3

InChI key

PSSCMQMQOXVMFG-UHFFFAOYSA-N

General description

3,5-Dichlorothioanisole is a meta-disubstituted thioanisole derivative. Its conformation has been studied based on the long-range spin-spin coupling constant and proximate proton-proton coupling constant, obtained from its NMR data.[1][2][3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanisms of long-range 13C, 13c spin-spin coupling in thioanisole and its derivatives. Conformational applications.
Schaefer T and Penner GH.
Canadian Journal of Chemistry, 66(5), 1229-1238 (1988)
R Kimura et al.
Toxicology and applied pharmacology, 78(2), 300-309 (1985-04-01)
The increases in the hepatic microsomal aminopyrine N-demethylase activity and in the content of cytochrome P-450 produced by m-dichlorobenzene (m-DCB) occurred after increases in the hepatic concentration of 3,5-dichlorophenyl methyl sulfone, a minor metabolite. The extent of increases in aminopyrine
Motion about the Csp 2-S bond in thioanisole and some derivatives by the J method.
Schaefer T and Baleja JD.
Canadian Journal of Chemistry, 64(7), 1326-1331 (1986)
The proximate coupling constant, 5 J (H, CH3), and the torsional mobility of the thiomethyl group in some thioanisole derivatives.
Schaefer T, et al.
Canadian Journal of Chemistry, 69(4), 620-624 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service