Skip to Content
MilliporeSigma
All Photos(1)

Documents

542741

Sigma-Aldrich

5-Bromothiophene-2-sulfonamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4BrNO2S2
CAS Number:
Molecular Weight:
242.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

138-142 °C (lit.)

SMILES string

NS(=O)(=O)c1ccc(Br)s1

InChI

1S/C4H4BrNO2S2/c5-3-1-2-4(9-3)10(6,7)8/h1-2H,(H2,6,7,8)

InChI key

WXJQQLDICAOBJB-UHFFFAOYSA-N

General description

5-Bromothiophene-2-sulfonamide can be synthesized by reacting 5-bromothiophene-2-sulfonyl fluoride with ammonium hydroxide in 2-methyltetrahydrofuran.

Application

5-Bromothiophene-2-sulfonamide may be used to synthesize:
  • 5-(phenylthio)thiophene-2-sulfonamide
  • 5-bromothiophene-2-sulfonyl acetamide
  • 5-arylthiophene-2-sulfonylacetamide derivatives

It may also be used in the preparation of the following 5-arylthiophene-2-sulfonamides:
  • 5-phenylthiophene-2-sulfonamide
  • 5-(3-cyno-5-(trifluoromethyl)phenyl)thiophene-2-sulfonamide
  • 5-(3,5-bis(trifluoromethyl)phenyl)thiophene-2-sulfonamide
  • 5′-chloro-2,2′-bithiophene-5-sulfonamide
  • 5′-methyl-2,2′-bithiophene-5-sulfonamide
  • 5-p-tolylthiophene-2-sulfonamide
  • 5-(4-methoxyphenyl)thiophene-2-sulfonamide
  • 5-(4-chlorophenyl)thiophene-2-sulfonamide
  • 5-(3,4-dichlorophenyl)thiophene-2-sulfonamide
  • 5-(3,5-dimethylphenyl)thiophene-2-sulfonamide
  • 5-(4-chloro-3-fluorophenyl)thiophene-2-sulfonamide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of an Acyl Sulfonamide Anti-Proliferative Agent, LY573636? Na?.
Yates MH, et al.
Organic Process Research & Development, 13(2), 255-262 (2009)
A facile synthesis of new 5-aryl-thiophenes bearing sulfonamide moiety via Pd (0)-catalyzed Suzuki-Miyaura cross coupling reactions and 5-bromothiophene-2-acetamide: As potent urease inhibitor, antibacterial agent and hemolytically active compounds.
Noreen M, et al.
Journal of Saudi Chemical Society (2014)
Mary M Mader et al.
Bioorganic & medicinal chemistry letters, 15(3), 617-620 (2005-01-25)
The anti-proliferative activity of acylated heterocyclic sulfonamides is described in Vascular Endothelial Growth Factor-dependent Human Umbilical Vascular Endothelial Cells (VEGF-HUVEC) and in HCT116 tumor cells in a soft agar diffusion assay.
I T Barnish et al.
Journal of medicinal chemistry, 24(8), 959-964 (1981-08-01)
A series of 5-(arylthio)-, 5-(arylsulfinyl)-, and 5-(arylsulfonyl)thiophene-2-sulfonamides is described and anticonvulsant activities are listed for the compounds. In most cases, the sulfones had the highest activity and the sulfides the least. Sulfones with 3- or 4-halo substituents generally had the
Mnaza Noreen et al.
Molecules (Basel, Switzerland), 20(11), 19914-19928 (2015-11-12)
A variety of novel 5-aryl thiophenes 4a-g containing sulphonylacetamide (sulfacetamide) groups were synthesized in appreciable yields via Pd[0] Suzuki cross coupling reactions. The structures of these newly synthesized compounds were determined using spectral data and elemental analysis. Density functional theory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service