About This Item
Recommended Products
assay
97%
refractive index
n20/D 1.5545 (lit.)
bp
217-218 °C (lit.)
density
1.459 g/mL at 25 °C (lit.)
SMILES string
Fc1ccc(Cl)cc1C(Cl)=O
InChI
1S/C7H3Cl2FO/c8-4-1-2-6(10)5(3-4)7(9)11/h1-3H
InChI key
VWGAATKTEAKOCO-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
212.0 °F - closed cup
flash_point_c
100 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service