Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

532533

Sigma-Aldrich

(S)-(−)-1,1,1-Trifluoro-2-octanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CH(OH)(CH2)5CH3
CAS Number:
Molecular Weight:
184.20
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.384 (lit.)

bp

170 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

SMILES string

CCCCCC[C@H](O)C(F)(F)F

InChI

1S/C8H15F3O/c1-2-3-4-5-6-7(12)8(9,10)11/h7,12H,2-6H2,1H3/t7-/m0/s1

InChI key

INAIBHXNHIEDAM-ZETCQYMHSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

163.9 °F - closed cup

flash_point_c

73.3 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service