Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

528773

Sigma-Aldrich

Dimethyl 5-methoxyisophthalate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CO2CH3)2
CAS Number:
Molecular Weight:
224.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

108-112 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1cc(OC)cc(c1)C(=O)OC

InChI

1S/C11H12O5/c1-14-9-5-7(10(12)15-2)4-8(6-9)11(13)16-3/h4-6H,1-3H3

InChI key

XZWYGKPSIBDYDY-UHFFFAOYSA-N

General description

Dimethyl 5-methoxyisophthalate can be prepared by reacting 5-methoxy isophthalic acid, 5-hydroxy isophthalic acid, K2CO3 and dimethyl sulphate.

Application

Dimethyl 5-methoxyisophthalate may be used as a reactant in the synthesis of 5-methoxy isophthalic acid.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura J McCormick et al.
CrystEngComm, 18(7), 1123-1132 (2016-03-29)
The synthesis and characterisation of five coordination polymers - Ni
Laura J McCormick et al.
Dalton transactions (Cambridge, England : 2003), 44(40), 17686-17695 (2015-09-24)
Solvothermal reaction of Zn(OAc)2 and 5-methoxy isophthalic acid (H2MeOip) in aqueous alcohols ROH (R = H, Me, Et, or (i)Pr) affords four different novel coordination polymers. Zn2(HMeOip)(MeOip)(OAc) (1) forms as a 1D 'ribbon of rings' polymer. Zn6(MeOip)4.5(HMeOip)(OH)2(H2O)2·5.5H2O (2) crystallises as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service