Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

525340

Sigma-Aldrich

4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CF3)NCS
CAS Number:
Molecular Weight:
237.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.5905 (lit.)

bp

248-251 °C (lit.)

density

1.47 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(ccc1Cl)N=C=S

InChI

1S/C8H3ClF3NS/c9-7-2-1-5(13-4-14)3-6(7)8(10,11)12/h1-3H

InChI key

AHFPRSSHNSGRCU-UHFFFAOYSA-N

General description

4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate is an aryl isothiocyanate. It can be synthesized from the corresponding aniline. Its freezing point is 308.15K. Enthalpy of vaporization of 4-chloro-3-(trifluoromethyl)phenyl isothiocyanate at boiling point (522.65K) is 46.065kjoule/mol.

Application

4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate may be used to synthesize aminotetrazole derivative.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals (2015)
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 548-548 (2014)
A highly efficient methodology for 5-methyl-3-aryl-2-thiooxazolidin-4-ones using lithium perchlorate in DIPEA mediated synthesis.
Khatik GL, et al.
Journal of Heterocyclic Chemistry, 47(3), 734-734 (2010)
Sara Van Poecke et al.
Bioorganic & medicinal chemistry, 19(24), 7603-7611 (2011-11-09)
We report on Mycobacterium tuberculosis thymidine monophosphate kinase (TMPKmt) inhibitory activities of a series of new 3'- and 5'-modified thymidine analogues including α- and β-derivatives. In addition, several analogues were synthesized in which the 4-oxygen was replaced by a more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service