Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

523364

Sigma-Aldrich

2-Bromo-3-nitropyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3BrN2O2
CAS Number:
Molecular Weight:
202.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

122-125 °C (lit.)

functional group

bromo
nitro

SMILES string

[O-][N+](=O)c1cccnc1Br

InChI

1S/C5H3BrN2O2/c6-5-4(8(9)10)2-1-3-7-5/h1-3H

InChI key

ZCCUFLITCDHRMG-UHFFFAOYSA-N

General description

2-Bromo-3-nitropyridine is a pyridine derivative. It can be synthesized from 3-nitropyridine-2-amine.[1]

application

2-Bromo-3-nitropyridine may be used in the synthesis of the following:
  • 3-nitropyridine-2-carbonitrile[1]
  • pyrrolo[3,2-b]pyridine[2]
  • 3-(hetero)arylated phenothiazines[3]
  • 7-anilino-6-azaindole-1-benznesulfonamides[4]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Condensed heteroaromatic ring systems. XII. Synthesis of indole derivatives from ethyl 2-bromocarbanilates.
Sakamoto T, et al.
Chemical & Pharmaceutical Bulletin, 35(5), 1823-1828 (1987)
Hsueh-Yun Lee et al.
Journal of medicinal chemistry, 56(20), 8008-8018 (2013-10-11)
Preliminary biological data on 7-anilino-6-azaindoles (8-11) suggested that hydrophobic substituents at C7 contribute to enhancement of antiproliferative activity. A novel series of 7-aryl-6-azaindole-1-benzenesulfonamides (12-22) were developed and showed improved cytotoxicity compared to ABT751 (5). The conversion of C7 phenyl rings
Yvonnick Loidreau et al.
European journal of medicinal chemistry, 58, 171-183 (2012-11-06)
A useful and rapid access to libraries of N-arylbenzo[b]thieno[3,2-d]pyrimidin-4-amines and their pyrido and pyrazino analogues was designed and optimized for the first time via microwave-accelerated condensation and Dimroth rearrangement of the starting anilines with N'-(2-cyanoaryl)-N,N-dimethylformimidamides obtained by reaction of thiophene
Synthesis and Electronic Properties of 3-Acceptor-Substituted and 3, 7-Bisacceptor-Substituted Phenothiazines.
Sailer M, et al.
European Journal of Organic Chemistry, 2006(2), 423-435 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service