Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

521809

Sigma-Aldrich

PSS-(4-(Chloromethyl)phenyl)-Heptacyclopentyl substituted

95%

Synonym(s):

1-(4-Chlorobenzyl)-3,5,7,9,11,13,15-heptacyclopentylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H69ClO12Si8
CAS Number:
Molecular Weight:
1026.13
MDL number:
UNSPSC Code:
12162002

assay

95%

mp

280 °C (dec.) (lit.)

SMILES string

Clc1ccc(C[Si]23O[Si]4(O[Si]5(O[Si](O2)(O[Si]6(O[Si](O3)(O[Si](O4)(O[Si](O5)(O6)C7CCCC7)C8CCCC8)C9CCCC9)C%10CCCC%10)C%11CCCC%11)C%12CCCC%12)C%13CCCC%13)cc1

InChI

1S/C42H69ClO12Si8/c43-35-31-29-34(30-32-35)33-56-44-57(36-15-1-2-16-36)47-60(39-21-7-8-22-39)49-58(45-56,37-17-3-4-18-37)51-62(41-25-11-12-26-41)52-59(46-56,38-19-5-6-20-38)50-61(48-57,40-23-9-10-24-40)54-63(53-60,55-62)42-27-13-14-28-42/h29-32,36-42H,1-28,33H2

Inchi Key

KDAMIASBEBBZQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in glass bottles

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service