Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

521779

Sigma-Aldrich

4′-Hydroxybutyranilide

97%

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$58.31

$58.31

List Price$83.30Save 30%
Web-Only Promotion

Available to ship onApril 14, 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$58.31

About This Item

Linear Formula:
CH3CH2CH2CONHC6H4OH
CAS Number:
Molecular Weight:
179.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$58.31

List Price$83.30Save 30%
Web-Only Promotion

Available to ship onApril 14, 2025Details


Request a Bulk Order

assay

97%

mp

137-141 °C (lit.)

functional group

amide

SMILES string

CCCC(=O)Nc1ccc(O)cc1

InChI

1S/C10H13NO2/c1-2-3-10(13)11-8-4-6-9(12)7-5-8/h4-7,12H,2-3H2,1H3,(H,11,13)

Inchi Key

KESXDDATSRRGAH-UHFFFAOYSA-N

General description

4′-Hydroxybutyranilide is an aromatic amide.

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service