Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

521337

Sigma-Aldrich

2-(Benzyloxy)phenylboronic acid

Synonym(s):

2-(Phenylmethoxy)benzeneboronic acid, 2-(Phenylmethyl)oxyphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4B(OH)2
CAS Number:
Molecular Weight:
228.05
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

mp

105-110 °C (lit.)

SMILES string

OB(O)c1ccccc1OCc2ccccc2

InChI

1S/C13H13BO3/c15-14(16)12-8-4-5-9-13(12)17-10-11-6-2-1-3-7-11/h1-9,15-16H,10H2

InChI key

MCAIDINWZOCYQK-UHFFFAOYSA-N

Application

Reactant for:
  • Palladium complex-catalyzed selective hydroxylation
  • Palladium(II)-catalyzed oxidative Heck reactions
  • Metal-free electrophilic fluorination
  • Suzuki-Miyaura cross-coupling reactions

Other Notes

Contains varying amounts of anhydride

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service