Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

520055

Sigma-Aldrich

4-Ethynylanisole

Synonym(s):

1-Ethynyl-4-methoxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4C≡CH
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352100

refractive index

n20/D 1.563 (lit.)

bp

87-91 °C/11 mmHg (lit.)

mp

28-29 °C (lit.)

density

1.019 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(cc1)C#C

InChI

1S/C9H8O/c1-3-8-4-6-9(10-2)7-5-8/h1,4-7H,2H3

InChI key

KBIAVTUACPKPFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Manufactured by CB Research and Development

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Chen et al.
Bioorganic & medicinal chemistry letters, 17(7), 1979-1983 (2007-02-16)
A library of potential antifungal triazole-modified beta-methoxyacrylate analogues was designed and synthesized via a Cu(I)-catalyzed 1,3-dipolar alkyne-azide coupling reaction or 'click chemistry'. Subsequent biological screening revealed that some compounds displayed low to moderate antifungal activity toward pathogenic fungi and low

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service