Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

519545

Sigma-Aldrich

tert-Dodecylmercaptan

Synonym(s):

tert-Dodecanethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H25SH
CAS Number:
Molecular Weight:
202.40
Beilstein/REAXYS Number:
1738382
EC Number:
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

availability

available only in EU

bp

227-248 °C (lit.)

density

0.86 g/mL at 20 °C (lit.)

SMILES string

SC(C(C(C(C)C)(C)C)(C)C)(C)C

InChI

1S/C12H26S/c1-9(2)10(3,4)11(5,6)12(7,8)13/h9,13H,1-8H3

InChI key

YAJYJWXEWKRTPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

203.0 °F - closed cup

flash_point_c

95 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service