Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

518425

Sigma-Aldrich

3-Bromo-5-fluoro-2-methoxyphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7BBrFO3
CAS Number:
Molecular Weight:
248.84
MDL number:
UNSPSC Code:
12352103

SMILES string

COc1c(Br)cc(F)cc1B(O)O

InChI

1S/C7H7BBrFO3/c1-13-7-5(8(11)12)2-4(10)3-6(7)9/h2-3,11-12H,1H3

InChI key

FUSHKJRKYBGAQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Contains varying amounts of anhydride

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service