Skip to Content
MilliporeSigma
All Photos(1)

Documents

516422

Sigma-Aldrich

4-Methoxy-3-nitrobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(NO2)CHO
CAS Number:
Molecular Weight:
181.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

97-100 °C (lit.)

SMILES string

COc1ccc(C=O)cc1[N+]([O-])=O

InChI

1S/C8H7NO4/c1-13-8-3-2-6(5-10)4-7(8)9(11)12/h2-5H,1H3

InChI key

YTCRQCGRYCKYNO-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua R Ring et al.
Acta crystallographica. Section C, Crystal structure communications, 63(Pt 7), o392-o394 (2007-07-05)
In the title compound, C(9)H(12)N(5)O(3)(+) x Cl(-), the cation is almost entirely planar. The imine double bond is exclusively in the E geometry.
Polymorphism in 4-methoxy-3-nitrobenzaldehyde.
Wishkerman S, et al.
Crystal Growth & Design, 6(6), 1366-1373 (2006)
Xiaojun Han et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(28), 7957-7964 (2007-07-06)
This work demonstrates the use of photocleavable cholesterol derivatives to create supported bilayer lipid membrane arrays on silica. The photocleavable cholesteryl tether is attached to the surface by using the reaction of an amine-functionalized self-assembled monolayer (SAM) and the N-hydroxysuccinimide-based
Tele nucleophilic aromatic substitutions in 1-nitro-3-and 1, 3-dinitro-5-trichloromethylbenzene, and 3-trichloromethylbenzonitrile. A new synthesis of the 1, 4-benzothiazine-3 (4H)-one ring system from 3-nitrobenzoic acid.
Giannopoulos T, et al.
Tetrahedron, 56(3), 447-453 (2000)
Synthesis, cytotoxic activity and docking studies of new 4-aza-podophyllotoxin derivatives.
Hatti I, et al.
Medicinal Chemistry Research, 24(8), 3305-3313 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service