Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

514594

Sigma-Aldrich

4′-Bromo-[1,1′-biphenyl]-4-ol acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C6H4C6H4Br
CAS Number:
Molecular Weight:
291.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Product 514594 is not currently sold in your country. Contact Technical Service

mp

130-134 °C (lit.)

SMILES string

CC(=O)Oc1ccc(cc1)-c2ccc(Br)cc2

InChI

1S/C14H11BrO2/c1-10(16)17-14-8-4-12(5-9-14)11-2-6-13(15)7-3-11/h2-9H,1H3

InChI key

PZPSICDSQWAIBR-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service