Skip to Content
MilliporeSigma
All Photos(1)

Documents

514403

Sigma-Aldrich

3-Ethoxy-2-methyl-2-cyclopenten-1-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC5H4(CH3)(=O)
CAS Number:
Molecular Weight:
140.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

bp

93-100 °C/0.5 mmHg (lit.)

mp

39-43 °C (lit.)

SMILES string

CCOC1=C(C)C(=O)CC1

InChI

1S/C8H12O2/c1-3-10-8-5-4-7(9)6(8)2/h3-5H2,1-2H3

InChI key

LNFQBEVBVGDZOJ-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service