Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

514322

Sigma-Aldrich

Bismuth(III) trifluoromethanesulfonate

Synonym(s):

Bi(OTf)3, Bismuth tris(trifluoromethanesulfonate), Bismuth(III) triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bi(OSO2CF3)3
CAS Number:
Molecular Weight:
656.19
MDL number:
Pricing and availability is not currently available.

mp

>300 °C (lit.)

SMILES string

[Bi+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Bi/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

NYENCOMLZDQKNH-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Displays very good catalyst activity in Mukaiyama aldol and Diels-Alder reactions.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Louer, M. et al.
Chemistry of Materials, 9, 3012-3012 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service