Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

514039

Sigma-Aldrich

2,3-Difluorophenylboronic acid

≥95%

Synonym(s):

2,3-Difluorobenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3B(OH)2
CAS Number:
Molecular Weight:
157.91
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

≥95%

mp

95 °C (dec.) (lit.)

functional group

fluoro

SMILES string

OB(O)c1cccc(F)c1F

InChI

1S/C6H5BF2O2/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3,10-11H

InChI key

SZYXKFKWFYUOGZ-UHFFFAOYSA-N

Application

Reactant for:
  • Suzuki-Miyaura cross-coupling reactions
  • Preparation of biologically and pharmacologically active molecules

Other Notes

Contains varying amounts of anhydride

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service