Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

513903

Sigma-Aldrich

Benzyl 4-hydroxy-1-piperidinecarboxylate

97%

Synonym(s):

1-(Benzyloxycarbonyl)-4-piperidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO3
CAS Number:
Molecular Weight:
235.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.543 (lit.)

bp

167 °C/0.2 mmHg (lit.)

density

1.554 g/mL at 25 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

OC1CCN(CC1)C(=O)OCc2ccccc2

InChI

1S/C13H17NO3/c15-12-6-8-14(9-7-12)13(16)17-10-11-4-2-1-3-5-11/h1-5,12,15H,6-10H2

InChI key

JKIUUDJOCYHIGY-UHFFFAOYSA-N

application

Reactant for synthesis of:
Orally bioavailable P2Y12 antagonists for inhibition of platelet aggregation
Piperidine derivatives
Molecular rods
Oxazolidinone-quinolone hybrids and their antibacterial activity
Cyclic prodrug of RGD peptidomimetic

Reactant for oxidation of alcohols

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service