513407
Luperox® DTA, Di-tert-amyl peroxide
97%
Synonym(s):
Di-tert-amyl peroxide
About This Item
assay
97%
reaction suitability
reagent type: oxidant
refractive index
n20/D 1.409 (lit.)
bp
146 °C (lit.)
density
0.818 g/mL at 25 °C (lit.)
SMILES string
CCC(C)(C)OOC(C)(C)CC
InChI
1S/C10H22O2/c1-7-9(3,4)11-12-10(5,6)8-2/h7-8H2,1-6H3
InChI key
JJRDRFZYKKFYMO-UHFFFAOYSA-N
Application
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 3 - Org. Perox. E - Skin Irrit. 2
Storage Class
5.2 - Organic peroxides and self-reacting hazardous materials
wgk_germany
WGK 2
flash_point_f
77.0 °F - closed cup
flash_point_c
25 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service