Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

512966

Sigma-Aldrich

Z-L-Prolinol

97%

Synonym(s):

N-Cbz-L-prolinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO3
CAS Number:
Molecular Weight:
235.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

97%

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.54 (lit.)

bp

160 °C/0.8 mmHg (lit.)

density

1.196 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC[C@@H]1CCCN1C(=O)OCc2ccccc2

InChI

1S/C13H17NO3/c15-9-12-7-4-8-14(12)13(16)17-10-11-5-2-1-3-6-11/h1-3,5-6,12,15H,4,7-10H2/t12-/m0/s1

InChI key

BJTNHGVCFWDNDP-LBPRGKRZSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service