511161
Ethyl 2-methyl-3-indoleacetate
98%
Synonym(s):
2-Methyl-1H-indole-3-acetic acid ethyl ester, NSC 289352
About This Item
Recommended Products
assay
98%
refractive index
n20/D 1.571 (lit.)
bp
195-196 °C/3.5 mmHg (lit.)
density
1.11 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)Cc1c(C)[nH]c2ccccc12
InChI
1S/C13H15NO2/c1-3-16-13(15)8-11-9(2)14-12-7-5-4-6-10(11)12/h4-7,14H,3,8H2,1-2H3
InChI key
SLEXJGHKKHQSDC-UHFFFAOYSA-N
General description
Application
- Reactant for preparation of hydroxamate derivatives as HDAC inhibitor with anticancer activity
- Reactant for stereoselective preparation of AG-041R, as a Potent Gastrin/CCK-B Receptor Antagonist
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service