Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

510831

Sigma-Aldrich

Poly(3-octylthiophene-2,5-diyl)

regiorandom

Synonym(s):

P3OT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18S)n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

mol wt

average Mn 22,000
average Mw 36,600

color

red

mp

198-211 °C

solubility

chloroform, methylene chloride, toluene, and THF: soluble

fluorescence

λex 419 nm; λem 550 nm in chloroform

OPV device performance

ITO/PEDOT:PSS/P3OT:PC61BM (1:2)/LiF/Al

  • Short-circuit current density (Jsc): 5.55 mA/cm2
  • Open-circuit voltage (Voc): 0.5 V
  • Fill Factor (FF): 0.33
  • Power Conversion Efficiency (PCE): 0.91 %

Looking for similar products? Visit Product Comparison Guide

General description

Poly(3-alkylthiophene)s exhibit thermo- and solvatochromism. Regiorandom poly(3-octylthiophene) shows relatively weak red PL in dilute solution. The emission is largely quenched in concentrated solutions and solid thin films.[1]1:1 (head-to-head):(head-to-tail) linkages of regioisomers.
Poly(3-octylthiophene-2,5-diyl) (P3OT) is an alkylthiophene based conductive polymer with high electroluminescence. It is soluble in most of the organic solvents and can be prepared by the electrochemical polymerization of 3-octylthiophene (3OT). It has hydrophobic properties and can be used as a transducer for ion selective electrodes.[2][3][4]

Application

Conducting polymer.
Rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials.
P3OT forms a nanocomposite with carbon nanotubes (CNTs) for devising a high performance semiconducting material.[5] It can also be used as a π-conjugating polymer for the fabrication of organic electronic based devices which include polymeric diodes, solar cells and photovoltaic modules.[6][7][8]
Used in organic field-effect transistors and in polymer-based solar cells.

Features and Benefits

Good processability, environmental stability and electroactivity.

Packaging

Packaged in glass bottles

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Semiconducting polymer diodes: Large size, low cost photodetectors with excellent visible-ultraviolet sensitivity
Yu, G and Pakbaz, K and Heeger, AJ
Applied Physics Letters, 64(25), 3422-3424 (1994)
Poly (3-octylthiophene) as solid contact for ion-selective electrodes: contradictions and possibilities
Jarvis JM, et al.
Journal of Solid State Electrochemistry, 20(11), 3033-3041 (2016)
Light-Emitting Polymers
Pei Q, et al.
Material Matters, 2(3), 26-26 (2007)
Single-wall carbon nanotube/conjugated polymer photovoltaic devices
Kymakis E and Amaratunga G
Applied Physics Letters, 80(1), 112-114 (2002)
Fabrication of a poly (3-octylthiophene-2, 5-diyl) electrochemiluminescence device assisted by perylene
Daimon T and Nihei E
Materials, 6(5), 1704-1717 (2013)

Articles

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service