Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

491225

Sigma-Aldrich

1-Ethyl-3-methylimidazolium chloride-d11

98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$1,534.50

$1,534.50

List Price$1,860.00Save 18%
Web-Only Promotion

Available to ship onApril 22, 2025Details


Request more information

Select a Size

Change View
100 MG
$1,534.50

About This Item

Empirical Formula (Hill Notation):
C6D11ClN2
CAS Number:
Molecular Weight:
157.69
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$1,534.50

List Price$1,860.00Save 18%
Web-Only Promotion

Available to ship onApril 22, 2025Details


Request more information

isotopic purity

98 atom % D

Quality Level

mp

77-79 °C (lit.)

mass shift

M+11

SMILES string

[Cl-].[2H]c1c([2H])[n+](c([2H])n1C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C6H11N2.ClH/c1-3-8-5-4-7(2)6-8;/h4-6H,3H2,1-2H3;1H/q+1;/p-1/i1D3,2D3,3D2,4D,5D,6D;

InChI key

BMQZYMYBQZGEEY-WQTMNAJCSA-M

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service