Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

478504

Sigma-Aldrich

3-Isocyanatobenzoyl chloride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCNC6H4COCl
CAS Number:
Molecular Weight:
181.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

refractive index

n20/D 1.5803 (lit.)

bp

91 °C/3 mmHg (lit.)

density

1.333 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)c1cccc(c1)N=C=O

InChI

1S/C8H4ClNO2/c9-8(12)6-2-1-3-7(4-6)10-5-11/h1-4H

InChI key

WBLVOWAHPPPNLF-UHFFFAOYSA-N

General description

3-Isocyanatobenzoyl chloride, also known as 2-isocyanato-1-methoxy-4-methylbenzene, is an aryl isocyanate.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymers containing the benzimidazole and the quinazolinedione unit in the main chain.
Iwakura Y, et al.
Macromolecular Chemistry and Physics, 176(1), 23-36 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service